Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-6-nitroquinoline, 97%

$39.32 - $131.84

Chemical Identifiers

Chemical Name or Material 5-Amino-6-nitroquinoline
CAS 35975-00-9
Molecular Formula C9H7N3O2
Formula Weight 189.17
MDL Number MFCD00006798
Beilstein 166360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0207803

  SDS
alfa aesar™
L0207803
1g Each for $39.32

AAL0207806

  SDS
alfa aesar™
L0207806
5g Each for $131.84
Description & Specifications

Specifications

Formula Weight 189.17
MDL Number MFCD00006798
Molecular Formula C9H7N3O2
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 35975-00-9
Assay 97%
Beilstein 166360
Melting Point ≈273°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-Amino-6-nitroquinoline
Quantity 1g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.