Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-4-bromo-1-methyl-1H-pyrazole, 98%

$57.57 - $177.80

Chemical Identifiers

Chemical Name or Material 5-Amino-4-bromo-1-methyl-1H-pyrazole
CAS 105675-85-2
Molecular Formula C4H6BrN3
Formula Weight 176.02
MDL Number MFCD00068157
Beilstein 7579192
Synonym 4-Bromo-2-methyl-1H-pyrazol-3-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1162003

  SDS
alfa aesar™
L1162003
1g Each for $177.80

AAL11620MD

  SDS
alfa aesar™
L11620MD
250mg Each for $57.57
Description & Specifications

Specifications

Formula Weight 176.02
MDL Number MFCD00068157
CAS 105675-85-2
Chemical Name or Material 5-Amino-4-bromo-1-methyl-1H-pyrazole
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C4H6BrN3
Synonym 4-Bromo-2-methyl-1H-pyrazol-3-amine
Melting Point 96° to 101°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Beilstein 7579192
Quantity 1g
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.