missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-4-bromo-1-methyl-1H-pyrazole, 98%

Manufacturer:  Alfa Aesar™ L1162003

 View more versions of this product

Catalog No. AAL1162003



Specifications

Specifications

5-Amino-4-bromo-1-methyl-1H-pyrazole
1g
C4H6BrN3
7579192
ODFDZOQJRPDQDF-UHFFFAOYSA-N
4-bromo-2-methylpyrazol-3-amine
600351
98%
96°C to 101°C
105675-85-2
MFCD00068157
4-bromo-1-methyl-1h-pyrazol-5-amine, 3-amino-4-bromo-2-methylpyrazole, 5-amino-4-bromo-1-methylpyrazole, 5-amino-4-bromo-1-methyl-1h-pyrazole, 1h-pyrazol-5-amine,4-bromo-1-methyl, 4-bromo-2-methyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 4-bromo-1-methyl, 4-bromo-1-methylpyrazole-5-ylamine, zlchem 1177, acmc-1c4z4
CN1C(=C(C=N1)Br)N
176.017
176.02

Chemical Identifiers

105675-85-2
176.017
ODFDZOQJRPDQDF-UHFFFAOYSA-N
600351
CN1C(=C(C=N1)Br)N
C4H6BrN3
MFCD00068157
4-bromo-1-methyl-1h-pyrazol-5-amine, 3-amino-4-bromo-2-methylpyrazole, 5-amino-4-bromo-1-methylpyrazole, 5-amino-4-bromo-1-methyl-1h-pyrazole, 1h-pyrazol-5-amine,4-bromo-1-methyl, 4-bromo-2-methyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 4-bromo-1-methyl, 4-bromo-1-methylpyrazole-5-ylamine, zlchem 1177, acmc-1c4z4
4-bromo-2-methylpyrazol-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents