Please login to your online account to display your discounted pricing

5-Amino-3-phenylisoxazole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Amino-3-phenylisoxazole
CAS 4369-55-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H8N2O
Formula Weight 160.18
MDL Number MFCD00464114
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC433400010

  SDS
thermo fisher scientific
AC433400010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC433400050

  SDS
thermo fisher scientific
AC433400050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 160.18
Assay ≥96.0%
Quantity 1g
Color Orange
Melting Point 105° to 114°C
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C9H8N2O
Assay Percent Range 97%
CAS 4369-55-5
MDL Number MFCD00464114
Chemical Name or Material 5-Amino-3-phenylisoxazole
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.