missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-3-phenylisoxazole, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC433400050

 View more versions of this product

Catalog No. AC433400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Amino-3-phenylisoxazole
Glass bottle
105°C to 114°C
4369-55-5
100.0
C9H8N2O
3-phenylisoxazol-5-amine, 5-amino-3-phenylisoxazole, 5-isoxazolamine, 3-phenyl, 3-phenyl-5-isoxazolamine, 3-phenylisoxazole-5-ylamine, isoxazole, 5-amino-3-phenyl, phenylisoxazolamine, pubchem15538, 3-phenylisoxazole-5-amine, acmc-209jv8
C1=CC=C(C=C1)C2=NOC(=C2)N
160.176
160.18
≥96.0%
Authentic
Orange
5g
96.0
97%
MFCD00464114
HLOHVVZZMMMDMM-UHFFFAOYSA-N
3-phenyl-1,2-oxazol-5-amine
261201
Crystalline Powder

Chemical Identifiers

4369-55-5
160.176
HLOHVVZZMMMDMM-UHFFFAOYSA-N
261201
C1=CC=C(C=C1)C2=NOC(=C2)N
C9H8N2O
MFCD00464114
3-phenylisoxazol-5-amine, 5-amino-3-phenylisoxazole, 5-isoxazolamine, 3-phenyl, 3-phenyl-5-isoxazolamine, 3-phenylisoxazole-5-ylamine, isoxazole, 5-amino-3-phenyl, phenylisoxazolamine, pubchem15538, 3-phenylisoxazole-5-amine, acmc-209jv8
3-phenyl-1,2-oxazol-5-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents