Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-3-phenylisoxazole, 97%

$137.26 - $482.16

Chemical Identifiers

Chemical Name or Material 5-Amino-3-phenylisoxazole
CAS 4369-55-5
Molecular Formula C9H8N2O
Formula Weight 160.17
MDL Number MFCD00464114
Synonym 3-Phenyl-5-isoxazolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2780103

  SDS
alfa aesar™
H2780103
1g Each for $137.26

AAH2780106

  SDS
alfa aesar™
H2780106
5g Each for $482.16
Description & Specifications

Specifications

Formula Weight 160.17
Quantity 1g
Health Hazard 1 Danger
Molecular Formula C9H8N2O
CAS 4369-55-5
Synonym 3-Phenyl-5-isoxazolamine
Assay 97%
Chemical Name or Material 5-Amino-3-phenylisoxazole
MDL Number MFCD00464114
Melting Point 108° to 110°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.