Learn More
5-Amino-3-methyl-1-phenyl-1H-pyrazole, ≥99%, Thermo Scientific Chemicals
$73.01 - $73.01
Chemical Identifiers
CAS | 1131-18-6 |
---|---|
Molecular Formula | C10H11N3 |
Molecular Weight (g/mol) | 173.219 |
MDL Number | MFCD00020727 |
InChI Key | FMKMKBLHMONXJM-UHFFFAOYSA-N |
Synonym | 3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl |
PubChem CID | 70801 |
IUPAC Name | 5-methyl-2-phenylpyrazol-3-amine |
SMILES | CC1=NN(C(=C1)N)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6360506
|
Thermo Scientific Chemicals
H6360506 |
5 g |
Each for $73.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1131-18-6 | |
173.219 | |
FMKMKBLHMONXJM-UHFFFAOYSA-N | |
70801 | |
CC1=NN(C(=C1)N)C2=CC=CC=C2 |
C10H11N3 | |
MFCD00020727 | |
3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl | |
5-methyl-2-phenylpyrazol-3-amine |
Specifications
1131-18-6 | |
Brown to Yellow | |
MFCD00020727 | |
140666 | |
FMKMKBLHMONXJM-UHFFFAOYSA-N | |
5-methyl-2-phenylpyrazol-3-amine | |
70801 | |
≥99% | |
5-Amino-3-methyl-1-phenyl-1H-pyrazole |
114°C to 117°C | |
C10H11N3 | |
5 g | |
3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl | |
CC1=NN(C(=C1)N)C2=CC=CC=C2 | |
173.219 | |
173.21 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 214-463-3
RTECSNumber : UQ4990000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only