missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-3-methyl-1-phenyl-1H-pyrazole, ≥99%

Manufacturer:  Alfa Aesar™ H6360514

 View more versions of this product

Catalog No. AAH6360514



Specifications

Specifications

5-Amino-3-methyl-1-phenyl-1H-pyrazole
114°C to 117°C
1131-18-6
MFCD00020727
3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
CC1=NN(C(=C1)N)C2=CC=CC=C2
173.219
173.21
≥99%
Brown to Yellow
25g
C10H11N3
140666
FMKMKBLHMONXJM-UHFFFAOYSA-N
5-methyl-2-phenylpyrazol-3-amine
70801
Solid

Chemical Identifiers

1131-18-6
173.219
FMKMKBLHMONXJM-UHFFFAOYSA-N
70801
CC1=NN(C(=C1)N)C2=CC=CC=C2
C10H11N3
MFCD00020727
3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
5-methyl-2-phenylpyrazol-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UQ4990000

TSCA : TSCA

SDS Documents