missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Amino-3-(4-methylphenyl)pyrazole, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 78597-54-3 |
---|---|
Molecular Formula | C10H11N3 |
Molecular Weight (g/mol) | 173.22 |
InChI Key | GVPFRVKDBZWRCZ-UHFFFAOYSA-N |
Synonym | 5-p-tolyl-1h-pyrazol-3-amine, 3-4-methylphenyl-1h-pyrazol-5-amine, 5-p-tolyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methylphenyl pyrazole, 3-amino-5-4-methylphenyl-1h-pyrazole, 3-amino-5-p-tolylpyrazole, 5-4-methylphenyl-1h-pyrazol-3-amine, 3-p-tolyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-4-methylphenyl, 5-4-methylphenyl-2h-pyrazol-3-amine |
PubChem CID | 736768 |
IUPAC Name | 5-(4-methylphenyl)-1H-pyrazol-3-amine |
SMILES | CC1=CC=C(C=C1)C2=CC(=NN2)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC438800050
|
Thermo Scientific Chemicals
438800050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
78597-54-3 | |
173.22 | |
5-p-tolyl-1h-pyrazol-3-amine, 3-4-methylphenyl-1h-pyrazol-5-amine, 5-p-tolyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methylphenyl pyrazole, 3-amino-5-4-methylphenyl-1h-pyrazole, 3-amino-5-p-tolylpyrazole, 5-4-methylphenyl-1h-pyrazol-3-amine, 3-p-tolyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-4-methylphenyl, 5-4-methylphenyl-2h-pyrazol-3-amine | |
5-(4-methylphenyl)-1H-pyrazol-3-amine |
C10H11N3 | |
GVPFRVKDBZWRCZ-UHFFFAOYSA-N | |
736768 | |
CC1=CC=C(C=C1)C2=CC(=NN2)N |
Specifications
78597-54-3 | |
100.0 | |
96% min. (HPLC) | |
5g | |
GVPFRVKDBZWRCZ-UHFFFAOYSA-N | |
5-(4-methylphenyl)-1H-pyrazol-3-amine | |
736768 | |
97% |
96.0 | |
Authentic | |
C10H11N3 | |
5-p-tolyl-1h-pyrazol-3-amine, 3-4-methylphenyl-1h-pyrazol-5-amine, 5-p-tolyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methylphenyl pyrazole, 3-amino-5-4-methylphenyl-1h-pyrazole, 3-amino-5-p-tolylpyrazole, 5-4-methylphenyl-1h-pyrazol-3-amine, 3-p-tolyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-4-methylphenyl, 5-4-methylphenyl-2h-pyrazol-3-amine | |
CC1=CC=C(C=C1)C2=CC(=NN2)N | |
173.22 | |
173.22 | |
5-Amino-3-(4-methylphenyl)pyrazole |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only