missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-3-(4-methylphenyl)pyrazole, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC438800010

 View more versions of this product

Catalog No. AC438800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Amino-3-(4-methylphenyl)pyrazole
96.0
White
96% min. (HPLC)
C10H11N3
1h-pyrazol-3-amine, 5-4-methylphenyl, 3-4-methylphenyl-1h-pyrazol-5-amine, 3-amino-5-4-methylphenyl-1h-pyrazole, 3-amino-5-p-tolylpyrazole, 3-p-tolyl-1h-pyrazol-5-amine, 5-4-methylphenyl-1h-pyrazol-3-amine, 5-4-methylphenyl-2h-pyrazol-3-amine, 5-amino-3-4-methylphenyl pyrazole, 5-p-tolyl-1h-pyrazol-3-amine, 5-p-tolyl-2h-pyrazol-3-ylamine
GVPFRVKDBZWRCZ-UHFFFAOYSA-N
5-(4-methylphenyl)-1H-pyrazol-3-amine
736768
Powder
78597-54-3
100.0
152.3°C to 153.3°C
Authentic
Glass Bottle
1g
CC1=CC=C(C=C1)C2=CC(=NN2)N
173.22
173.22
97%

Chemical Identifiers

78597-54-3
173.22
1h-pyrazol-3-amine, 5-4-methylphenyl, 3-4-methylphenyl-1h-pyrazol-5-amine, 3-amino-5-4-methylphenyl-1h-pyrazole, 3-amino-5-p-tolylpyrazole, 3-p-tolyl-1h-pyrazol-5-amine, 5-4-methylphenyl-1h-pyrazol-3-amine, 5-4-methylphenyl-2h-pyrazol-3-amine, 5-amino-3-4-methylphenyl pyrazole, 5-p-tolyl-1h-pyrazol-3-amine, 5-p-tolyl-2h-pyrazol-3-ylamine
5-(4-methylphenyl)-1H-pyrazol-3-amine
C10H11N3
GVPFRVKDBZWRCZ-UHFFFAOYSA-N
736768
CC1=CC=C(C=C1)C2=CC(=NN2)N
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only