Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-3,4-dimethylisoxazole, 97%

$53.57 - $163.16

Chemical Identifiers

Chemical Name or Material 5-Amino-3,4-dimethylisoxazole
CAS 19947-75-2
Molecular Formula C5H8N2O
Formula Weight 112.13
MDL Number MFCD00046081
Beilstein 108546
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1219104

 
alfa aesar™
L1219104
2g Each for $53.57

AAL1219109

 
alfa aesar™
L1219109
10g Each for $163.16
Description & Specifications

Specifications

Formula Weight 112.13
Chemical Name or Material 5-Amino-3,4-dimethylisoxazole
Assay 97%
Quantity 2g
Molecular Formula C5H8N2O
MDL Number MFCD00046081
CAS 19947-75-2
Beilstein 108546
Melting Point 118° to 120°C