missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Maybridge

Chemical Identifiers

CAS 42754-62-1
Molecular Formula C10H7ClN4
Molecular Weight (g/mol) 218.644
MDL Number MFCD02677738
InChI Key DWFJNGUSRCQFDM-UHFFFAOYSA-N
Synonym 5-amino-3-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 3-amino-5-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 3-amino-5-4-chlorophenyl-2h-pyrazole-4-carbonitrile, 5-4-chlorophenyl-3-aminopyrazole-4-carbonitrile, 5-amino-3-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile,3-amino-5-4-chlorophenyl
PubChem CID 2776122
IUPAC Name 3-amino-5-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
SMILES C1=CC(=CC=C1C2=C(C(=NN2)N)C#N)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC00044CB SDS View Documents Fisher Scientific Company
CC00044CB
250mg Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

42754-62-1
218.644
DWFJNGUSRCQFDM-UHFFFAOYSA-N
2776122
C1=CC(=CC=C1C2=C(C(=NN2)N)C#N)Cl
C10H7ClN4
MFCD02677738
5-amino-3-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 3-amino-5-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 3-amino-5-4-chlorophenyl-2h-pyrazole-4-carbonitrile, 5-4-chlorophenyl-3-aminopyrazole-4-carbonitrile, 5-amino-3-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile,3-amino-5-4-chlorophenyl
3-amino-5-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile

Specifications

Amber Glass Bottle
42754-62-1
100.0
C10H7ClN4
5-amino-3-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 3-amino-5-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 3-amino-5-4-chlorophenyl-2h-pyrazole-4-carbonitrile, 5-4-chlorophenyl-3-aminopyrazole-4-carbonitrile, 5-amino-3-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile,3-amino-5-4-chlorophenyl
C1=CC(=CC=C1C2=C(C(=NN2)N)C#N)Cl
218.644
218.65
5-Amino-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
250mg
96.5
97%
MFCD02677738
DWFJNGUSRCQFDM-UHFFFAOYSA-N
3-amino-5-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
2776122
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.