Please login to your online account to display your discounted pricing

5-Amino-2-(trifluoromethyl)benzimidazole, 96%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Amino-2-(trifluoromethyl)benzimidazole
Assay Percent Range 96%
Molecular Formula C8H6F3N3
Formula Weight 201.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442340050

 
thermo fisher scientific
AC442340050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442340010

 
thermo fisher scientific
AC442340010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.15
Quantity 5g
Chemical Name or Material 5-Amino-2-(trifluoromethyl)benzimidazole
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C8H6F3N3
CAS 3671-66-7