missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-2-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444560010

 View more versions of this product

Catalog No. AC444560010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Amino-2-methylphenylboronic acid pinacol ester
83°C to 84°C
882670-69-1
C13H20BNO2
HPNBWHDRLCVZFV-UHFFFAOYSA-N
16086089
97%
Glass Bottle
1g
97%
4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 5-amino-2-methyl-phenylboronic acid pinacol ester, 5-amino-2-methylphenyl boronic acid pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester, 5-amino-2-methylphenylboronic acid, pinacol ester, benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, hpnbwhdrlcvzfv-uhfffaoysa-n
233.12
233.12

Chemical Identifiers

882670-69-1
233.12
4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 5-amino-2-methyl-phenylboronic acid pinacol ester, 5-amino-2-methylphenyl boronic acid pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester, 5-amino-2-methylphenylboronic acid, pinacol ester, benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, hpnbwhdrlcvzfv-uhfffaoysa-n
C13H20BNO2
HPNBWHDRLCVZFV-UHFFFAOYSA-N
16086089
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents