missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-2-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 882670-69-1
Molecular Formula C13H20BNO2
Molecular Weight (g/mol) 233.12
InChI Key HPNBWHDRLCVZFV-UHFFFAOYSA-N
Synonym 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-amino-2-methylphenylboronic acid, pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, hpnbwhdrlcvzfv-uhfffaoysa-n, 5-amino-2-methyl-phenylboronic acid pinacol ester, 5-amino-2-methylphenyl boronic acid pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester
PubChem CID 16086089
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444560010 SDS View Documents Acros Organics
AC444560010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444560050 SDS View Documents Acros Organics
AC444560050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

882670-69-1
233.12
4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-amino-2-methylphenylboronic acid, pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, hpnbwhdrlcvzfv-uhfffaoysa-n, 5-amino-2-methyl-phenylboronic acid pinacol ester, 5-amino-2-methylphenyl boronic acid pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester
C13H20BNO2
HPNBWHDRLCVZFV-UHFFFAOYSA-N
16086089

Specifications

1g
97%
4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-amino-2-methylphenylboronic acid, pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, hpnbwhdrlcvzfv-uhfffaoysa-n, 5-amino-2-methyl-phenylboronic acid pinacol ester, 5-amino-2-methylphenyl boronic acid pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester
233.12
233.12
882670-69-1
C13H20BNO2
HPNBWHDRLCVZFV-UHFFFAOYSA-N
16086089
5-Amino-2-methylphenylboronic acid pinacol ester
Safety and Handling

Safety and Handling

Warning