Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-methylindole, 97%

$48.07 - $718.73

Chemical Identifiers

Chemical Name or Material 5-Amino-2-methylindole
CAS 7570-49-2
Molecular Formula C9H10N2
Formula Weight 145.19
MDL Number MFCD02093426
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2169403

  SDS
alfa aesar™
B2169403
1g Each for $48.07

AAB2169406

  SDS
alfa aesar™
B2169406
5g Each for $182.32

AAB2169414

  SDS
alfa aesar™
B2169414
25g Each for $718.73
Description & Specifications

Specifications

Formula Weight 145.19
Molecular Formula C9H10N2
Chemical Name or Material 5-Amino-2-methylindole
Health Hazard 1 Danger
Assay 97%
MDL Number MFCD02093426
Recommended Storage 2° to 8°C
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
CAS 7570-49-2
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.

2° to 8°C