missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-2-methylbenzonitrile, 97%, ACROS Organics™

Chemical Identifiers

CAS 50670-64-9
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
InChI Key YDZVQWCVKXYGIU-UHFFFAOYSA-N
Synonym 2-methyl-5-aminobenzonitrile, 3-amino-6-methylbenzonitrile, 3-cyano-4-methylaniline, 4-amino-2-cyanotoluene, 5-amino-2-methyl-benzonitrile, 5-amino-2-methylbenzenecarbonitrile, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, benzonitrile, 5-amino-2-methyl
PubChem CID 2735365
IUPAC Name 5-amino-2-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439850050
View Documents
Thermo Fisher Scientific
AC439850050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439850250
SDS View Documents
Thermo Fisher Scientific
AC439850250
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

50670-64-9
132.17
2-methyl-5-aminobenzonitrile, 3-amino-6-methylbenzonitrile, 3-cyano-4-methylaniline, 4-amino-2-cyanotoluene, 5-amino-2-methyl-benzonitrile, 5-amino-2-methylbenzenecarbonitrile, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, benzonitrile, 5-amino-2-methyl
5-amino-2-methylbenzonitrile
C8H8N2
YDZVQWCVKXYGIU-UHFFFAOYSA-N
2735365
CC1=C(C=C(C=C1)N)C#N

Specifications

Authentic
90.9°C to 91.7°C
50670-64-9
100.0
C8H8N2
YDZVQWCVKXYGIU-UHFFFAOYSA-N
5-amino-2-methylbenzonitrile
2735365
Powder
5-Amino-2-methylbenzonitrile
Yellow to Brown
5g
96.0
96% min. (GC)
2-methyl-5-aminobenzonitrile, 3-amino-6-methylbenzonitrile, 3-cyano-4-methylaniline, 4-amino-2-cyanotoluene, 5-amino-2-methyl-benzonitrile, 5-amino-2-methylbenzenecarbonitrile, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, benzonitrile, 5-amino-2-methyl
CC1=C(C=C(C=C1)N)C#N
132.17
132.17
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Warning