missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Amino-2-methylbenzonitrile, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439850250
Specifications
5-Amino-2-methylbenzonitrile | |
Yellow to Brown | |
25g | |
96.0 | |
96% min. (GC) | |
2-methyl-5-aminobenzonitrile, 3-amino-6-methylbenzonitrile, 3-cyano-4-methylaniline, 4-amino-2-cyanotoluene, 5-amino-2-methyl-benzonitrile, 5-amino-2-methylbenzenecarbonitrile, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, benzonitrile, 5-amino-2-methyl | |
CC1=C(C=C(C=C1)N)C#N | |
132.17 | |
132.17 | |
97% |
Authentic | |
90.9°C to 91.7°C | |
50670-64-9 | |
100.0 | |
C8H8N2 | |
YDZVQWCVKXYGIU-UHFFFAOYSA-N | |
5-amino-2-methylbenzonitrile | |
2735365 | |
Powder |
Chemical Identifiers
50670-64-9 | |
132.17 | |
2-methyl-5-aminobenzonitrile, 3-amino-6-methylbenzonitrile, 3-cyano-4-methylaniline, 4-amino-2-cyanotoluene, 5-amino-2-methyl-benzonitrile, 5-amino-2-methylbenzenecarbonitrile, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, benzonitrile, 5-amino-2-methyl | |
5-amino-2-methylbenzonitrile |
C8H8N2 | |
YDZVQWCVKXYGIU-UHFFFAOYSA-N | |
2735365 | |
CC1=C(C=C(C=C1)N)C#N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
GHS Signal Word: Warning