missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-methylbenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H6142206

 View more versions of this product

Catalog No. AAH6142206



Specifications

Specifications

5-Amino-2-methylbenzonitrile
5g
C8H8N2
UN3439
YDZVQWCVKXYGIU-UHFFFAOYSA-N
5-amino-2-methylbenzonitrile
2735365
Solid
88°C to 91°C
50670-64-9
MFCD00017608
2-methyl-5-aminobenzonitrile, 3-amino-6-methylbenzonitrile, 3-cyano-4-methylaniline, 4-amino-2-cyanotoluene, 5-amino-2-methyl-benzonitrile, 5-amino-2-methylbenzenecarbonitrile, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, benzonitrile, 5-amino-2-methyl
CC1=C(C=C(C=C1)N)C#N
132.166
132.16
97%

Chemical Identifiers

50670-64-9
132.166
YDZVQWCVKXYGIU-UHFFFAOYSA-N
2735365
CC1=C(C=C(C=C1)N)C#N
C8H8N2
MFCD00017608
2-methyl-5-aminobenzonitrile, 3-amino-6-methylbenzonitrile, 3-cyano-4-methylaniline, 4-amino-2-cyanotoluene, 5-amino-2-methyl-benzonitrile, 5-amino-2-methylbenzenecarbonitrile, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, benzonitrile, 5-amino-2-methyl
5-amino-2-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents