Learn More
Alfa Aesar™ 5-Amino-2-methylbenzonitrile, 97%
Manufacturer: Alfa Aesar™ H6142214
Specifications
5-Amino-2-methylbenzonitrile | |
25g | |
C8H8N2 | |
UN3439 | |
YDZVQWCVKXYGIU-UHFFFAOYSA-N | |
5-amino-2-methylbenzonitrile | |
2735365 | |
Solid |
88°C to 91°C | |
50670-64-9 | |
MFCD00017608 | |
2-methyl-5-aminobenzonitrile | |
CC1=C(C=C(C=C1)N)C#N | |
132.166 | |
132.16 | |
97% |
Chemical Identifiers
50670-64-9 | |
132.166 | |
YDZVQWCVKXYGIU-UHFFFAOYSA-N | |
2735365 | |
CC1=C(C=C(C=C1)N)C#N |
C8H8N2 | |
MFCD00017608 | |
2-methyl-5-aminobenzonitrile | |
5-amino-2-methylbenzonitrile |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.