missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-methylbenzoic acid, 97%

$38.09 - $153.12

Chemical Identifiers

CAS 2855-08-5
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD06208351
InChI Key XGCKOSFYXBAPQM-UHFFFAOYSA-N
Synonym neohexyl chloride, 3,3-dimethylbutyl chloride, butane, 1-chloro-3,3-dimethyl, 1-chloro-3,3-dimethyl-butane, acmc-1clex, tert-butylchloroethylene, ksc561m9t, butane,1-chloro-3,3-dimethyl, 1-chloro-3,3-dimethylbutane 10g, 3,3-paragraph signthorn 1/4 x>>u-1-ae paragraph sign inverted exclamation markie
PubChem CID 76111
IUPAC Name 1-chloro-3,3-dimethylbutane
SMILES CC(C)(C)CCCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6007803 View Documents Alfa Aesar™
H6007803
1g Each for $38.09
 
AAH6007806 View Documents Alfa Aesar™
H6007806
5g Each for $153.12
 
Specifications

Chemical Identifiers

2855-08-5
120.62
XGCKOSFYXBAPQM-UHFFFAOYSA-N
76111
CC(C)(C)CCCl
C6H13Cl
MFCD06208351
neohexyl chloride, 3,3-dimethylbutyl chloride, butane, 1-chloro-3,3-dimethyl, 1-chloro-3,3-dimethyl-butane, acmc-1clex, tert-butylchloroethylene, ksc561m9t, butane,1-chloro-3,3-dimethyl, 1-chloro-3,3-dimethylbutane 10g, 3,3-paragraph signthorn 1/4 x>>u-1-ae paragraph sign inverted exclamation markie
1-chloro-3,3-dimethylbutane

Specifications

194°C to 199°C
2840-04-2
MFCD06208351
FSXVZWAWYKMFMX-UHFFFAOYSA-N
5-amino-2-methylbenzoic acid
10374614
97%
1g
C8H9NO2
5-amino-2-methyl-benzoic acid, 5-amino-2-methyl benzoic acid, 2-methyl-5-aminobenzoic acid, 5-amino-o-toluic acid, 5-amino-2-methylbenzoicacid, benzoic acid, 5-amino-2-methyl, pubchem4980, 3-amino-6-methylbenzoic acid, ksc494q3t, 2-methyl-5-amino benzoic acid
CC1=C(C=C(C=C1)N)C(=O)O
151.17
151.16
5-Amino-2-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C