missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5′-Amino-2′-methylacetanilide 98.0+%, TCI America™

$156.13 - $534.67

Chemical Identifiers

CAS 5434-30-0
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
MDL Number MFCD00047859
InChI Key UAZGSMMESOKKQZ-UHFFFAOYSA-N
Synonym 2-acetylamino-4-aminotoluene, 2-acetylamino-p-toluidine, 2-methyl-5-aminoacetaniline, 3-acetamido-4-methylaniline, 3-acetylamino-p-toluidine, 5'-amino-2'-methylacetanilide, acetamide, n-5-amino-2-methylphenyl, n-3-amino-6-methylphenyl acetamide, n-5-amino-2-methylphenyl acetamide, n1-5-amino-2-methylphenyl acetamide
PubChem CID 79492
IUPAC Name N-(5-amino-2-methylphenyl)acetamide
SMILES CC1=C(C=C(C=C1)N)NC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A27241G
View Documents
Tci America
A27241G
1g
Each for $156.13
 
A27245G
View Documents
Tci America
A27245G
5g
Each for $534.67
 
Description

Description

Specifications

Chemical Identifiers

5434-30-0
164.208
UAZGSMMESOKKQZ-UHFFFAOYSA-N
79492
CC1=C(C=C(C=C1)N)NC(=O)C
C9H12N2O
MFCD00047859
2-acetylamino-4-aminotoluene, 2-acetylamino-p-toluidine, 2-methyl-5-aminoacetaniline, 3-acetamido-4-methylaniline, 3-acetylamino-p-toluidine, 5'-amino-2'-methylacetanilide, acetamide, n-5-amino-2-methylphenyl, n-3-amino-6-methylphenyl acetamide, n-5-amino-2-methylphenyl acetamide, n1-5-amino-2-methylphenyl acetamide
N-(5-amino-2-methylphenyl)acetamide

Specifications

White-Yellow
1g
C9H12N2O
2-acetylamino-4-aminotoluene, 2-acetylamino-p-toluidine, 2-methyl-5-aminoacetaniline, 3-acetamido-4-methylaniline, 3-acetylamino-p-toluidine, 5'-amino-2'-methylacetanilide, acetamide, n-5-amino-2-methylphenyl, n-3-amino-6-methylphenyl acetamide, n-5-amino-2-methylphenyl acetamide, n1-5-amino-2-methylphenyl acetamide
CC1=C(C=C(C=C1)N)NC(=O)C
164.208
164.21
≥98.0% (HPLC,T)
144°C
5434-30-0
MFCD00047859
UAZGSMMESOKKQZ-UHFFFAOYSA-N
N-(5-amino-2-methylphenyl)acetamide
79492
Crystal-Powder at 20°C
5′-Amino-2′-methylacetanilide
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0270

TSCA : Yes

SDS
Product Certifications