missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-2-methyl-1,3-benzoxazole, 97%, ACROS Organics™

Chemical Identifiers

CAS 72745-76-7
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.17
InChI Key FECYTFWPNCBIHC-UHFFFAOYSA-N
Synonym 2-methyl-5-aminobenzoxazole, 2-methyl-benzooxazol-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 2-methylbenzo d oxazol-5-amine, 2-methylbenzoxazol-5-yl amine, 2-methylbenzoxazole-5-ylamine, 5-amino-2-methyl-1,3-benzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine, 2-methyl, 5-benzoxazolamine,2-methyl
PubChem CID 737920
IUPAC Name 2-methyl-1,3-benzoxazol-5-amine
SMILES CC1=NC2=C(O1)C=CC(=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC437920010
SDS View Documents
Thermo Fisher Scientific
AC437920010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437920050
SDS View Documents
Thermo Fisher Scientific
AC437920050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

72745-76-7
148.17
2-methyl-5-aminobenzoxazole, 2-methyl-benzooxazol-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 2-methylbenzo d oxazol-5-amine, 2-methylbenzoxazol-5-yl amine, 2-methylbenzoxazole-5-ylamine, 5-amino-2-methyl-1,3-benzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine, 2-methyl, 5-benzoxazolamine,2-methyl
2-methyl-1,3-benzoxazol-5-amine
C8H8N2O
FECYTFWPNCBIHC-UHFFFAOYSA-N
737920
CC1=NC2=C(O1)C=CC(=C2)N

Specifications

Authentic
White to Brown
1g
96.0
96% min. (HPLC)
2-methyl-5-aminobenzoxazole, 2-methyl-benzooxazol-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 2-methylbenzo d oxazol-5-amine, 2-methylbenzoxazol-5-yl amine, 2-methylbenzoxazole-5-ylamine, 5-amino-2-methyl-1,3-benzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine, 2-methyl, 5-benzoxazolamine,2-methyl
CC1=NC2=C(O1)C=CC(=C2)N
148.17
148.17
97%
Glass Bottle
72°C to 74.6°C
72745-76-7
100.0
C8H8N2O
FECYTFWPNCBIHC-UHFFFAOYSA-N
2-methyl-1,3-benzoxazol-5-amine
737920
Crystalline Powder
5-Amino-2-methyl-1, 3-benzoxazole
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face p

GHS Signal Word: Warning

SDS