missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Amino-2-methyl-1,3-benzoxazole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437920050
Specifications
5-Amino-2-methyl-1, 3-benzoxazole | |
Glass bottle | |
72°C to 74.6°C | |
72745-76-7 | |
100.0 | |
C8H8N2O | |
FECYTFWPNCBIHC-UHFFFAOYSA-N | |
2-methyl-1,3-benzoxazol-5-amine | |
737920 | |
Crystalline Powder |
Authentic | |
White to Brown | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-methyl-5-aminobenzoxazole, 2-methyl-benzooxazol-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 2-methylbenzo d oxazol-5-amine, 2-methylbenzoxazol-5-yl amine, 2-methylbenzoxazole-5-ylamine, 5-amino-2-methyl-1,3-benzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine, 2-methyl, 5-benzoxazolamine,2-methyl | |
CC1=NC2=C(O1)C=CC(=C2)N | |
148.17 | |
148.17 | |
97% |
Chemical Identifiers
72745-76-7 | |
148.17 | |
2-methyl-5-aminobenzoxazole, 2-methyl-benzooxazol-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 2-methylbenzo d oxazol-5-amine, 2-methylbenzoxazol-5-yl amine, 2-methylbenzoxazole-5-ylamine, 5-amino-2-methyl-1,3-benzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine, 2-methyl, 5-benzoxazolamine,2-methyl | |
2-methyl-1,3-benzoxazol-5-amine |
C8H8N2O | |
FECYTFWPNCBIHC-UHFFFAOYSA-N | |
737920 | |
CC1=NC2=C(O1)C=CC(=C2)N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face p
GHS Signal Word: Warning