Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-methoxypyridine, 98%

$68.35 - $212.42

Chemical Identifiers

Chemical Name or Material 5-Amino-2-methoxypyridine
CAS 6628-77-9
Molecular Formula C6H8N2O
Formula Weight 124.14
MDL Number MFCD00006264
Synonym 6-Methoxy-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2752806

  SDS
alfa aesar™
H2752806
5g Each for $68.35

AAH2752814

  SDS
alfa aesar™
H2752814
25g Each for $212.42
Description & Specifications

Specifications

Formula Weight 124.14
Chemical Name or Material 5-Amino-2-methoxypyridine
Assay 98%
Molecular Formula C6H8N2O
Melting Point 29° to 31°C
Boiling Point 85° to 90°C (1mmHg)
MDL Number MFCD00006264
CAS 6628-77-9
Quantity 5g
Flash Point >110°C (230°F)
Synonym 6-Methoxy-3-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number US1836000
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C