Please login to your online account to display your discounted pricing

5-Amino-2-methoxyphenol, 98%, ACROS Organics™


Quantity 10g

Manufacturer: thermo fisher scientific  33491-0100

 View more versions of this product

Catalog No. AC334910100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Color Gray-brown to dark brown
Physical Form Crystalline Powder
Formula Weight 139.15
Quantity 10g
EINECS Number 216-873-8
Melting Point 129° to 133°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 5-Amino-2-methoxyphenol
Molecular Formula C7H9NO2
CAS 1687-53-2
Assay Percent Range 98%
MDL Number MFCD00010222
Name Note 98%
Linear Formula C6H3NH(OH)OCH3
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Amino-2-methoxyphenol
Name Note 98%
CAS 1687-53-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H9NO2
Linear Formula C6H3NH(OH)OCH3
Formula Weight 139.15
MDL Number MFCD00010222
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.

SDS & Safety Documents