missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-2-methoxyphenol, 98%, ACROS Organics™

Chemical Identifiers

CAS 1687-53-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H9NO2
Linear Formula C6H3NH(OH)OCH3
Formula Weight 139.15
MDL Number MFCD00010222
Chemical Name or Material 5-Amino-2-methoxyphenol, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC334910100 SDS Thermo Fisher Scientific
33491-0100
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1687-53-2
100.0
C7H9NO2
139.15
5-Amino-2-methoxyphenol, 98%
97.5
98%
C6H3NH(OH)OCH3
MFCD00010222
≥97.5%

Specifications

Gray-brown to dark brown
139.15
129° to 133°C
Glass bottle
1687-53-2
MFCD00010222
97.5
5-Amino-2-methoxyphenol, 98%
Crystalline Powder
10g
100.0
C7H9NO2
98%
C6H3NH(OH)OCH3
Authentic
≥97.5%