Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-mercaptobenzimidazole, 96%

$97.95 - $286.28

Chemical Identifiers

Chemical Name or Material 5-Amino-2-mercaptobenzimidazole
CAS 2818-66-8
Molecular Formula C7H7N3S
Formula Weight 165.22
MDL Number MFCD00022671
Synonym 5-Amino-1,3-dihydro-2H-benzimidazole-2-thione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2699914

  SDS
alfa aesar™
H2699914
25g Each for $97.95

AAH2699922

  SDS
alfa aesar™
H2699922
100g Each for $286.28
Description & Specifications

Specifications

Formula Weight 165.22
Chemical Name or Material 5-Amino-2-mercaptobenzimidazole
TSCA TSCA
Assay 96%
Molecular Formula C7H7N3S
Melting Point 240° to 244°C
MDL Number MFCD00022671
CAS 2818-66-8
Quantity 25g
Synonym 5-Amino-1,3-dihydro-2H-benzimidazole-2-thione
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.