Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-fluoropyridine, 97+%

Chemical Identifiers

Chemical Name or Material 5-Amino-2-fluoropyridine
CAS 1827-27-6
Molecular Formula C5H5FN2
Formula Weight 112.11
MDL Number MFCD01632180
UN Number UN2811
Synonym 6-Fluoro-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3087203

 
alfa aesar™
H3087203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3087206

 
alfa aesar™
H3087206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.11
Chemical Name or Material 5-Amino-2-fluoropyridine
Assay ≥97%
Molecular Formula C5H5FN2
Melting Point 88° to 91°C
MDL Number MFCD01632180
CAS 1827-27-6
UN Number UN2811
Quantity 1g
Odor Characteristic
Synonym 6-Fluoro-3-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.