Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-fluorobenzoic acid, 99%

$100.51 - $100.51

Chemical Identifiers

Chemical Name or Material 5-Amino-2-fluorobenzoic acid
CAS 56741-33-4
Molecular Formula C7H6FNO2
Formula Weight 155.13
MDL Number MFCD00077449
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26162ME

  SDS
alfa aesar™
H26162ME
500mg Each for $100.51
Description & Specifications

Specifications

Formula Weight 155.13
Chemical Name or Material 5-Amino-2-fluorobenzoic acid
Assay 99%
Molecular Formula C7H6FNO2
Melting Point 190° to 192°C
MDL Number MFCD00077449
CAS 56741-33-4
Quantity 500mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.