missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-fluoro-4-methylbenzene boronic acid pinacol ester, 96%

Chemical Identifiers

CAS 1817801-55-0
Molecular Formula C13H19BFNO2
Molecular Weight (g/mol) 251.11
InChI Key MONFQILBYDTFKM-UHFFFAOYSA-N
Synonym 4-fluoro-2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62402MD View Documents Alfa Aesar™
H62402MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6240203 View Documents Alfa Aesar™
H6240203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1817801-55-0
251.11
4-fluoro-2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
C13H19BFNO2
MONFQILBYDTFKM-UHFFFAOYSA-N

Specifications

250mg
C13H19BFNO2
MONFQILBYDTFKM-UHFFFAOYSA-N
251.1
96%
1817801-55-0
4-fluoro-2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
251.11
Solid
5-Amino-2-fluoro-4-methylbenzene boronic acid pinacol ester