missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-fluoro-3-methylpyridine, 95%

$51.20 - $606.05

Chemical Identifiers

CAS 186593-48-6
Molecular Formula C6H7FN2
Molecular Weight (g/mol) 126.134
MDL Number MFCD03095088
InChI Key KADQKKYTJHXBSL-UHFFFAOYSA-N
Synonym 5-amino-2-fluoro-3-picoline, 2-fluoro-3-methyl-5-aminopyridine, 5-amino-2-fluoro-3-methylpyridine, 2-fluoro-5-amino-3-picoline, 6-fluoro-5-methyl-3-pyridinamine, 3-pyridinamine, 6-fluoro-5-methyl, 6-fluoro-5-methyl-3-pyridylamine, 5-amino-3-methyl-2-fluoropyridine, 3-pyridinamine, 6-fluoro-5-methyl-9ci, pubchem1258
PubChem CID 21952691
IUPAC Name 6-fluoro-5-methylpyridin-3-amine
SMILES CC1=CC(=CN=C1F)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64854MD View Documents Alfa Aesar™
H64854MD
250mg Each for $51.20
 
AAH6485403 View Documents Alfa Aesar™
H6485403
1g Each for $151.76
 
AAH6485406 View Documents Alfa Aesar™
H6485406
5g Each for $606.05
 
Specifications

Chemical Identifiers

186593-48-6
126.134
KADQKKYTJHXBSL-UHFFFAOYSA-N
21952691
CC1=CC(=CN=C1F)N
C6H7FN2
MFCD03095088
5-amino-2-fluoro-3-picoline, 2-fluoro-3-methyl-5-aminopyridine, 5-amino-2-fluoro-3-methylpyridine, 2-fluoro-5-amino-3-picoline, 6-fluoro-5-methyl-3-pyridinamine, 3-pyridinamine, 6-fluoro-5-methyl, 6-fluoro-5-methyl-3-pyridylamine, 5-amino-3-methyl-2-fluoropyridine, 3-pyridinamine, 6-fluoro-5-methyl-9ci, pubchem1258
6-fluoro-5-methylpyridin-3-amine

Specifications

250mg
C6H7FN2
13574091
KADQKKYTJHXBSL-UHFFFAOYSA-N
6-fluoro-5-methylpyridin-3-amine
21952691
Solid
5-Amino-2-fluoro-3-methylpyridine
186593-48-6
MFCD03095088
5-amino-2-fluoro-3-picoline, 2-fluoro-3-methyl-5-aminopyridine, 5-amino-2-fluoro-3-methylpyridine, 2-fluoro-5-amino-3-picoline, 6-fluoro-5-methyl-3-pyridinamine, 3-pyridinamine, 6-fluoro-5-methyl, 6-fluoro-5-methyl-3-pyridylamine, 5-amino-3-methyl-2-fluoropyridine, 3-pyridinamine, 6-fluoro-5-methyl-9ci, pubchem1258
CC1=CC(=CN=C1F)N
126.134
126.13
95%