Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-cyanopyridine, 96%

Chemical Identifiers

Chemical Name or Material 5-Amino-2-cyanopyridine
CAS 55338-73-3
Molecular Formula C6H5N3
Formula Weight 119.13
MDL Number MFCD03093956
UN Number UN3439
Synonym 5-Aminopicolinonitrile; 5-Aminopyridine-2-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2896014

  SDS
alfa aesar™
H2896014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2896022

  SDS
alfa aesar™
H2896022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 119.13
Synonym 5-Aminopicolinonitrile; 5-Aminopyridine-2-carbonitrile
Health Hazard 3 GHS P Statement
P280f-P305+P351+P338-P309-P310
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 148° to 152°C
Health Hazard 2 GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Molecular Formula C6H5N3
UN Number UN3439
MDL Number MFCD03093956
CAS 55338-73-3
Chemical Name or Material 5-Amino-2-cyanopyridine
Quantity 25g
Assay 96%

Applications
Silver(I)-assisted electrophilic aromatic iodination of commercially available 5-aminopicolinonitrile 2 gave the mono-iodinated intermediate 3 in 79% yield. In mild and efficient DBU-catalyzed amidation of cyanoacetates activation of the α-carboxyl group allowed condensation with 5-amino-2-cyanopyridine.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280f-P305+P351+P338-P309-P310
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.