missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Amino-2-chloropyrimidine, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 56621-90-0 |
---|---|
Molecular Formula | C4H4ClN3 |
Molecular Weight (g/mol) | 129.55 |
InChI Key | DZBKIOJXVOECRA-UHFFFAOYSA-N |
Synonym | 5-amino-2-chloropyrimidine, 2-chloro-pyrimidin-5-ylamine, 2-chloro-5-aminopyrimidine, 5-pyrimidinamine, 2-chloro, 2-chloropyrimidine-5-ylamine, pubchem6963, acmc-1axfh, 2-chloropyrimidin-5-ylamine, 5-pyrimidinamine,2-chloro, 5-amino-2-chloro pyrimidine |
PubChem CID | 12215993 |
IUPAC Name | 2-chloropyrimidin-5-amine |
SMILES | C1=C(C=NC(=N1)Cl)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC434320010
|
Thermo Scientific Chemicals
434320010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC434320050
|
Thermo Scientific Chemicals
434320050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
56621-90-0 | |
129.55 | |
5-amino-2-chloropyrimidine, 2-chloro-pyrimidin-5-ylamine, 2-chloro-5-aminopyrimidine, 5-pyrimidinamine, 2-chloro, 2-chloropyrimidine-5-ylamine, pubchem6963, acmc-1axfh, 2-chloropyrimidin-5-ylamine, 5-pyrimidinamine,2-chloro, 5-amino-2-chloro pyrimidine | |
2-chloropyrimidin-5-amine |
C4H4ClN3 | |
DZBKIOJXVOECRA-UHFFFAOYSA-N | |
12215993 | |
C1=C(C=NC(=N1)Cl)N |
Specifications
56621-90-0 | |
100.0 | |
94% min. (GC) | |
C4H4ClN3 | |
5-amino-2-chloropyrimidine, 2-chloro-pyrimidin-5-ylamine, 2-chloro-5-aminopyrimidine, 5-pyrimidinamine, 2-chloro, 2-chloropyrimidine-5-ylamine, pubchem6963, acmc-1axfh, 2-chloropyrimidin-5-ylamine, 5-pyrimidinamine,2-chloro, 5-amino-2-chloro pyrimidine | |
C1=C(C=NC(=N1)Cl)N | |
129.55 | |
129.55 | |
5-Amino-2-chloropyrimidine |
94.0 | |
Authentic | |
Glass bottle | |
1g | |
DZBKIOJXVOECRA-UHFFFAOYSA-N | |
2-chloropyrimidin-5-amine | |
12215993 | |
95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co
GHS Signal Word: Warning
RUO â Research Use Only