Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-chloropyridine, 98%

$76.50 - $76.50

Chemical Identifiers

Chemical Name or Material 5-Amino-2-chloropyridine
CAS 5350-93-6
Molecular Formula C5H5ClN2
Formula Weight 128.56
MDL Number MFCD00006243
Beilstein 108891
Synonym 6-Chloro-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2533603

 
alfa aesar™
B2533603
1g Each for $76.50
Description & Specifications

Specifications

Formula Weight 128.56
Chemical Name or Material 5-Amino-2-chloropyridine
Melting Point 81° to 84°C
Quantity 1g
Odor Unpleasant
Assay 98%
Molecular Formula C5H5ClN2
MDL Number MFCD00006243
CAS 5350-93-6
Beilstein 108891
Synonym 6-Chloro-3-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.