missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-chlorophenol, 98%

$68.00 - $986.96

Chemical Identifiers

CAS 6358-06-1
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD01707858
InChI Key JSCNCRWPXOTDDZ-UHFFFAOYSA-N
Synonym 2-chloro-5-aminophenol, 4-chloro-3-hydroxyaniline, phenol, 5-amino-2-chloro, 5-amino-2-chloro-phenol, 3-amino-6-chlorophenol, pubchem21902, 3-hydroxy-4-chloroaniline, 4-chloro-3-hydroxy aniline, acmc-1b64d, ksc494c4r
PubChem CID 80676
IUPAC Name 5-amino-2-chlorophenol
SMILES C1=CC(=C(C=C1N)O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6637106 View Documents Alfa Aesar™
H6637106
5g Each for $68.00
 
AAH6637114 View Documents Alfa Aesar™
H6637114
25g Each for $268.32
 
AAH6637122 View Documents Alfa Aesar™
H6637122
100g Each for $986.96
 
Specifications

Chemical Identifiers

6358-06-1
143.57
JSCNCRWPXOTDDZ-UHFFFAOYSA-N
80676
C1=CC(=C(C=C1N)O)Cl
C6H6ClNO
MFCD01707858
2-chloro-5-aminophenol, 4-chloro-3-hydroxyaniline, phenol, 5-amino-2-chloro, 5-amino-2-chloro-phenol, 3-amino-6-chlorophenol, pubchem21902, 3-hydroxy-4-chloroaniline, 4-chloro-3-hydroxy aniline, acmc-1b64d, ksc494c4r
5-amino-2-chlorophenol

Specifications

Light Sensitive
160°C
6358-08-3
MFCD01707858
2081092
MHAFRUMLQZZSIN-UHFFFAOYSA-N
2-amino-4-chloro-6-nitrophenol
4348294
Powder
5-Amino-2-chlorophenol
Gray
5g
C6H5ClN2O3
UN2811
unii-13d00yq1u0, 6-amino-4-chloro-2-nitrophenol, 2-amino-4-chloro-6-nitro-phenol, 2-amino-4-chloro-6-nitro phenol, phenol, 2-amino-4-chloro-6-nitro, acmc-2097jj, 4-phenylethyl isothiocyanate, 4-chlor-6-nitro-2-aminophenol, 5-chloro-2-hydroxy-3-nitroaniline, phenol,2-amino-4-chloro-6-nitro
C1=C(C=C(C(=C1[N+](=O)[O-])O)N)Cl
188.57
143.57
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : SJ5720000

storageNote1 : 2° to 8°C