missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Amino-2-chlorobenzotrifluoride, 99%, ACROS Organics™
$274.55 - $274.55
Chemical Identifiers
CAS | 320-51-4 |
---|---|
Molecular Formula | C7H5ClF3N |
Molecular Weight (g/mol) | 195.57 |
MDL Number | MFCD00007846 |
InChI Key | ASPDJZINBYYZRU-UHFFFAOYSA-N |
Synonym | 2-chloro-5-aminebenzotrifluoride, 2-chloro-5-aminobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-3-trifluoromethyl aniline, 4-chloro-3-trifluoromethyl benzenamine, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 5-amino-2-chlorobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl |
PubChem CID | 67574 |
ChEBI | CHEBI:59245 |
IUPAC Name | 4-chloro-3-(trifluoromethyl)aniline |
SMILES | C1=CC(=C(C=C1N)C(F)(F)F)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC103370050
|
Thermo Fisher Scientific
AC103370050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC103370250
|
Thermo Fisher Scientific
103370250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC103371000
|
Thermo Fisher Scientific
103371000 |
100g | Glass bottle |
Each for $274.55
|
|
Chemical Identifiers
320-51-4 | |
195.57 | |
ASPDJZINBYYZRU-UHFFFAOYSA-N | |
67574 | |
4-chloro-3-(trifluoromethyl)aniline |
C7H5ClF3N | |
MFCD00007846 | |
2-chloro-5-aminebenzotrifluoride, 2-chloro-5-aminobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-3-trifluoromethyl aniline, 4-chloro-3-trifluoromethyl benzenamine, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 5-amino-2-chlorobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl | |
CHEBI:59245 | |
C1=CC(=C(C=C1N)C(F)(F)F)Cl |
Specifications
134°C | |
Glass bottle | |
Orange or Pink to White | |
5g | |
98.5 | |
98.5% min. (GC) | |
H2NC6H3(Cl)CF3 | |
2-chloro-5-aminebenzotrifluoride, 2-chloro-5-aminobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-3-trifluoromethyl aniline, 4-chloro-3-trifluoromethyl benzenamine, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 5-amino-2-chlorobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl | |
ASPDJZINBYYZRU-UHFFFAOYSA-N | |
4-chloro-3-(trifluoromethyl)aniline | |
67574 | |
195.57 | |
99% |
Authentic | |
85.0°C (3.0 mmHg) | |
34.0°C to 38.0°C | |
320-51-4 | |
100.0 | |
C7H5ClF3N | |
MFCD00007846 | |
Solubility in water: negligible | |
C1=CC(=C(C=C1N)C(F)(F)F)Cl | |
195.57 | |
CHEBI:59245 | |
Crystalline Powder or Low Melting Solid | |
5-Amino-2-chlorobenzotrifluoride, 99% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
GHS Signal Word: Warning
EINECSNumber : 206-277-6
TSCA : TSCA