missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-chloro-4-methylpyridine, 98%

Manufacturer:  Alfa Aesar™ H6111506

 View more versions of this product

Catalog No. AAH6111506



Description

Description

Applications
5-Amino-2-chloro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

5-Amino-2-chloro-4-methylpyridine
5g
C6H7ClN2
3-amino-6-chloro-4-methyl-pyridine
PLSMBLIGZJWLEJ-UHFFFAOYSA-N
6-chloro-4-methylpyridin-3-amine
2734425
Solid
Orange
66909-38-4
MFCD03427661
Slightly soluble in water.
CC1=CC(=NC=C1N)Cl
142.586
142.59
98%

Chemical Identifiers

66909-38-4
142.586
PLSMBLIGZJWLEJ-UHFFFAOYSA-N
2734425
CC1=CC(=NC=C1N)Cl
C6H7ClN2
MFCD03427661
3-amino-6-chloro-4-methyl-pyridine
6-chloro-4-methylpyridin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

Documents