missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-2-chloro-3-picoline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC447630010

 View more versions of this product

Catalog No. AC447630010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Amino-2-chloro-3-picoline
90°C to 92°C
38186-82-2
C6H7ClN2
VSBISZPNLZFTPG-UHFFFAOYSA-N
6-chloro-5-methylpyridin-3-amine
12295607
97%
Glass Bottle
1g
96% min. (GC)
2-chloro-3-methyl-5-aminopyridine, 3-pyridinamine, 6-chloro-5-methyl, 5-amino-2-chloro-3-methylpyridine, 5-amino-2-chloro-3-picoline, 5-methyl-6-chloropyridine-3-amine, 6-chloro-5-methyl-3-pyridinamine, 6-chloro-5-methyl-3-pyridylamine, 6-chloro-5-methyl-pyridin-3-ylamine, 6-chloro-5-methylpyridine-3-amine, pubchem1152
CC1=CC(=CN=C1Cl)N
142.59
142.59

Chemical Identifiers

38186-82-2
142.59
2-chloro-3-methyl-5-aminopyridine, 3-pyridinamine, 6-chloro-5-methyl, 5-amino-2-chloro-3-methylpyridine, 5-amino-2-chloro-3-picoline, 5-methyl-6-chloropyridine-3-amine, 6-chloro-5-methyl-3-pyridinamine, 6-chloro-5-methyl-3-pyridylamine, 6-chloro-5-methyl-pyridin-3-ylamine, 6-chloro-5-methylpyridine-3-amine, pubchem1152
6-chloro-5-methylpyridin-3-amine
C6H7ClN2
VSBISZPNLZFTPG-UHFFFAOYSA-N
12295607
CC1=CC(=CN=C1Cl)N
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents