Please login to your online account to display your discounted pricing

5-Amino-2-chloro-3-picoline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Amino-2-chloro-3-picoline
Assay Percent Range 97%
Molecular Formula C6H7ClN2
Formula Weight 142.59
Synonym 5-Amino-2-chloro-3-methylpyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC447630050

 
thermo fisher scientific
AC447630050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC447630010

 
thermo fisher scientific
AC447630010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.59
Quantity 5g
Chemical Name or Material 5-Amino-2-chloro-3-picoline
Assay Percent Range 97%
Molecular Formula C6H7ClN2
CAS 38186-82-2
Synonym 5-Amino-2-chloro-3-methylpyridine