Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-bromopyridine, 97%

$56.04 - $680.36

Chemical Identifiers

Chemical Name or Material 5-Amino-2-bromopyridine
CAS 13534-97-9
Molecular Formula C5H5BrN2
Formula Weight 173.01
MDL Number MFCD01646116
Beilstein 109102
Synonym 6-Bromo-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2000803

 
alfa aesar™
L2000803
1g Each for $56.04

AAL2000806

 
alfa aesar™
L2000806
5g Each for $175.47

AAL2000814

 
alfa aesar™
L2000814
25g Each for $680.36
Description & Specifications

Specifications

Formula Weight 173.01
Assay 97%
Molecular Formula C5H5BrN2
MDL Number MFCD01646116
CAS 13534-97-9
Synonym 6-Bromo-3-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-Amino-2-bromopyridine
Melting Point 75° to 79°C
Quantity 1g
Beilstein 109102
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.