missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-bromo-3-methylpyridine, 98%

$91.91 - $365.49

Chemical Identifiers

CAS 38186-83-3
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD03095085
InChI Key IEONGQZSAUAAKH-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-3-picoline, 5-amino-2-bromo-3-methylpyridine, 3-pyridinamine, 6-bromo-5-methyl, 6-bromo-5-methyl-3-pyridylamine, 6-bromo-5-methyl-3-aminopyridine, pubchem1129, ksc985c1p, 2-bromo-3-methyl-5-aminopyridine, 2-bromo-5-amino-3-methylpyridine, 6-bromo-5-methyl-pyridin-3-ylamine
PubChem CID 12362446
IUPAC Name 6-bromo-5-methylpyridin-3-amine
SMILES CC1=CC(=CN=C1Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6613406 View Documents Alfa Aesar™
H6613406
5g Each for $91.91
 
AAH6613414 View Documents Alfa Aesar™
H6613414
25g Each for $365.49
 
Specifications

Chemical Identifiers

38186-83-3
187.04
IEONGQZSAUAAKH-UHFFFAOYSA-N
12362446
CC1=CC(=CN=C1Br)N
C6H7BrN2
MFCD03095085
5-amino-2-bromo-3-picoline, 5-amino-2-bromo-3-methylpyridine, 3-pyridinamine, 6-bromo-5-methyl, 6-bromo-5-methyl-3-pyridylamine, 6-bromo-5-methyl-3-aminopyridine, pubchem1129, ksc985c1p, 2-bromo-3-methyl-5-aminopyridine, 2-bromo-5-amino-3-methylpyridine, 6-bromo-5-methyl-pyridin-3-ylamine
6-bromo-5-methylpyridin-3-amine

Specifications

Gray
5g
C6H7BrN2
UN2811
5-amino-2-bromo-3-picoline, 5-amino-2-bromo-3-methylpyridine, 3-pyridinamine, 6-bromo-5-methyl, 6-bromo-5-methyl-3-pyridylamine, 6-bromo-5-methyl-3-aminopyridine, pubchem1129, ksc985c1p, 2-bromo-3-methyl-5-aminopyridine, 2-bromo-5-amino-3-methylpyridine, 6-bromo-5-methyl-pyridin-3-ylamine
CC1=CC(=CN=C1Br)N
187.04
187.04
98%
97°C to 100°C
38186-83-3
MFCD03095085
471832
IEONGQZSAUAAKH-UHFFFAOYSA-N
6-bromo-5-methylpyridin-3-amine
12362446
Solid
5-Amino-2-bromo-3-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C