missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2,4-dichloropyridine, 97%

$106.26 - $337.73

Chemical Identifiers

CAS 7321-93-9
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00234054
InChI Key FBGVTWONYOCYGA-UHFFFAOYSA-N
Synonym 5-amino-2,4-dichloropyridine, 4,6-dichloro-pyridin-3-ylamine, 3-pyridinamine, 4,6-dichloro, 4,6-dichloro-3-pyridinamine, 4,6-dichloropyridin-3-ylamine, pubchem17045, acmc-209opx, 2,4-dichloro-5-aminopyridine, 4,6-dichloropyridine-3-amine, 3-pyridinamine,4,6-dichloro
PubChem CID 595729
IUPAC Name 4,6-dichloropyridin-3-amine
SMILES C1=C(C(=CN=C1Cl)N)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66282MD View Documents Alfa Aesar™
H66282MD
250mg Each for $106.26
 
AAH6628203 View Documents Alfa Aesar™
H6628203
1g Each for $337.73
 
Specifications

Chemical Identifiers

7321-93-9
163.001
FBGVTWONYOCYGA-UHFFFAOYSA-N
595729
C1=C(C(=CN=C1Cl)N)Cl
C5H4Cl2N2
MFCD00234054
5-amino-2,4-dichloropyridine, 4,6-dichloro-pyridin-3-ylamine, 3-pyridinamine, 4,6-dichloro, 4,6-dichloro-3-pyridinamine, 4,6-dichloropyridin-3-ylamine, pubchem17045, acmc-209opx, 2,4-dichloro-5-aminopyridine, 4,6-dichloropyridine-3-amine, 3-pyridinamine,4,6-dichloro
4,6-dichloropyridin-3-amine

Specifications

Brown
7321-93-9
MFCD00234054
119452
FBGVTWONYOCYGA-UHFFFAOYSA-N
4,6-dichloropyridin-3-amine
595729
Powder
5-Amino-2,4-dichloropyridine
250mg
C5H4Cl2N2
UN2811
5-amino-2,4-dichloropyridine, 4,6-dichloro-pyridin-3-ylamine, 3-pyridinamine, 4,6-dichloro, 4,6-dichloro-3-pyridinamine, 4,6-dichloropyridin-3-ylamine, pubchem17045, acmc-209opx, 2,4-dichloro-5-aminopyridine, 4,6-dichloropyridine-3-amine, 3-pyridinamine,4,6-dichloro
C1=C(C(=CN=C1Cl)N)Cl
163.001
163
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : -30° to -10°C