Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2,4-dichlorophenol, 97%

Chemical Identifiers

Chemical Name or Material 5-Amino-2,4-dichlorophenol
CAS 39489-79-7
Formula Weight 178.01
MDL Number MFCD00186187
UN Number UN2811
Beilstein 2717129
Synonym 2,4-Dichloro-5-hydroxyaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6675303

 
alfa aesar™
H6675303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6675306

 
alfa aesar™
H6675306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66753MD

 
alfa aesar™
H66753MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 133° to 137°C
Formula Weight 178.01
Chemical Name or Material 5-Amino-2,4-dichlorophenol
Assay 97%
MDL Number MFCD00186187
CAS 39489-79-7
Synonym 2,4-Dichloro-5-hydroxyaniline
Physical Form Solid
Quantity 1g
UN Number UN2811
Beilstein 2717129
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C