Learn More
5-Amino-2,2-difluoro-1,3-benzodioxole, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2433603
Description
5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Light sensitive. Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
5-Amino-2,2-difluoro-1,3-benzodioxole | |
95°C to 97°C (12 mmHg) | |
C7H5F2NO2 | |
MFCD00190144 | |
UN2810 | |
Light sensitive | |
Not miscible or difficult to mix in water. | |
C1=CC2=C(C=C1N)OC(O2)(F)F | |
173.119 | |
173.12 |
1544-85-0 | |
Recognizable | |
1.499 | |
1 g | |
1343593 | |
2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine | |
CVYQRDKVWVBOFP-UHFFFAOYSA-N | |
2,2-difluoro-1,3-benzodioxol-5-amine | |
2736893 | |
≥97% |
Chemical Identifiers
1544-85-0 | |
173.119 | |
CVYQRDKVWVBOFP-UHFFFAOYSA-N | |
2736893 | |
C1=CC2=C(C=C1N)OC(O2)(F)F |
C7H5F2NO2 | |
MFCD00190144 | |
2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine | |
2,2-difluoro-1,3-benzodioxol-5-amine |
Safety and Handling
GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H301-H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only