Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-2-(2,4-dichlorophenoxy)pyridine, 99%

$109.85 - $382.72

Chemical Identifiers

Chemical Name or Material 5-Amino-2-(2,4-dichlorophenoxy)pyridine
CAS 85686-48-2
Molecular Formula C11H8Cl2N2O
Formula Weight 255.11
MDL Number MFCD00052643
Synonym 2,4-Dichlorophenoxy-5-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5068303

  SDS
alfa aesar™
H5068303
1g Each for $382.72

AAH50683MD

  SDS
alfa aesar™
H50683MD
250mg Each for $109.85
Description & Specifications

Specifications

Formula Weight 255.11
Assay 99%
CAS 85686-48-2
Molecular Formula C11H8Cl2N2O
Synonym 2,4-Dichlorophenoxy-5-pyridinamine
Quantity 1g
MDL Number MFCD00052643
Chemical Name or Material 5-Amino-2-(2,4-dichlorophenoxy)pyridine