Learn More
5-Amino-1H-tetrazole monohydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2401814
Description
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
Solubility
Soluble in water, ethanol (1.36%).
Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
Specifications
5-Amino-1H-tetrazole monohydrate | |
∼203°C (decomposition) | |
CH5N5O | |
25 g | |
108872 | |
Soluble in water,ethanol (1.36%). | |
C1(=NNN=N1)N.O | |
103.085 | |
103.08 (85.06 Anhydrous) |
15454-54-3 | |
3.7 | |
MFCD00149327 | |
UN1325 | |
1h-tetrazol-5-amine hydrate, 5-amino-1h-tetrazole monohydrate, 5-aminotetrazole monohydrate, 5-amino-1h-tetrazole hydrate, 5-aminotetrazole hydrate, 1h-1,2,3,4-tetrazol-5-amine hydrate, 2h-1,2,3,4-tetrazol-5-amine hydrate, tetrazol-5-amine, 5-aminotetrazole.h2o, 5-aminotetrazole h2o | |
JVSMPWHQUPKRNV-UHFFFAOYSA-N | |
2H-tetrazol-5-amine;hydrate | |
12211273 | |
99% |
Chemical Identifiers
15454-54-3 | |
103.085 | |
JVSMPWHQUPKRNV-UHFFFAOYSA-N | |
12211273 | |
C1(=NNN=N1)N.O |
CH5N5O | |
MFCD00149327 | |
1h-tetrazol-5-amine hydrate, 5-amino-1h-tetrazole monohydrate, 5-aminotetrazole monohydrate, 5-amino-1h-tetrazole hydrate, 5-aminotetrazole hydrate, 1h-1,2,3,4-tetrazol-5-amine hydrate, 2h-1,2,3,4-tetrazol-5-amine hydrate, tetrazol-5-amine, 5-aminotetrazole.h2o, 5-aminotetrazole h2o | |
2H-tetrazol-5-amine;hydrate |
Safety and Handling
GHS H Statement
H240-H228-H315-H319-H335
Heating may cause an explosion.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 224-581-7
RTECSNumber : XF7465000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only