missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-1H-benzimidazole-2-thiol, 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C7H7N3S
Formula Weight 165.22
Chemical Name or Material 5-Amino-1H-benzimidazole-2-thiol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC34641EE SDS Fisher Scientific Company
AC34641EE
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC34641EA SDS Fisher Scientific Company
AC34641EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
C7H7N3S
165.22
5-Amino-1H-benzimidazole-2-thiol

Specifications

165.22
2818-66-8
97%
50g
C7H7N3S
5-Amino-1H-benzimidazole-2-thiol