Please login to your online account to display your discounted pricing

5-Amino-1H-benzimidazole-2-thiol, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  AC34641EA

 View more versions of this product

Catalog No. AC34641EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 165.22
Quantity 10g
Chemical Name or Material 5-Amino-1H-benzimidazole-2-thiol
Assay Percent Range 97%
Molecular Formula C7H7N3S
CAS 2818-66-8

Specifications

Chemical Name or Material 5-Amino-1H-benzimidazole-2-thiol
Assay Percent Range 97%
Molecular Formula C7H7N3S
Formula Weight 165.22