Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile, 98%

Chemical Identifiers

Chemical Name or Material 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile
CAS 5334-43-0
Molecular Formula C10H8N4
Formula Weight 184.2
MDL Number MFCD00020730
UN Number UN3439
Beilstein 161503
Synonym 5-Amino-4-cyano-1-phenyl-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2067903

 
alfa aesar™
B2067903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2067906

 
alfa aesar™
B2067906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 184.2
Melting Point ≈138°C (dec.)
Quantity 1g
CAS 5334-43-0
Synonym 5-Amino-4-cyano-1-phenyl-1H-pyrazole
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Chemical Name or Material 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile
Assay 98%
Molecular Formula C10H8N4
MDL Number MFCD00020730
UN Number UN3439
Beilstein 161503
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.